Ethyl-5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

076628-500MG 076628-1G
102547-172EA 803.08 USD
102547-172 102547-174
Ethyl-5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylate ≥95%
Ethyl-5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₈F₆N₂O₂
MW: 338.21 g/mol
MDL Number: MFCD22373660
CAS Number: 1379526-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR