Methyl-2-cyclopentyl-2-hydroxyphenylacetate ≥95%
Supplier: Matrix Scientific
075682-1G
075682-5G
075682-10G
102542-506EA
613.85
USD
102542-506
102542-504
102542-508
Methyl-2-cyclopentyl-2-hydroxyphenylacetate ≥95%
Methyl-2-cyclopentyl-2-hydroxyphenylacetate
Hazard: Irritant
Formula:
C₁₄H₁₈O₃ |
MDL Number:
MFCD22194314 CAS Number: 1373253-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...