5-Chloro-2-(difluoromethyl)pyridine ≥95%
Supplier: Matrix Scientific
071610-1G
102520-428EA
638.46
USD
102520-428
5-Chloro-2-(difluoromethyl)pyridine ≥95%
5-Chloro-2-(difluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₄ClF₂N |
MDL Number:
MFCD22199314 CAS Number: 1374659-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...