2,6-Dichloro-3-(difluoromethyl)-5-fluoropyridine ≥97%

Supplier: Matrix Scientific

071600-1G
102520-412EA 606.15 USD
102520-412
2,6-Dichloro-3-(difluoromethyl)-5-fluoropyridine ≥97%
2,6-Dichloro-3-(difluoromethyl)-5-fluoropyridine

Hazard: Irritant

Formula: C₆H₂Cl₂F₃N
MDL Number: MFCD22199308
CAS Number: 1374659-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR