3,5-Dichloro-2-(difluoromethyl)pyridine ≥97%

071598-1G
102520-408EA 438.51 USD
102520-408
3,5-Dichloro-2-(difluoromethyl)pyridine ≥97%
3,5-Dichloro-2-(difluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃Cl₂F₂N
MDL Number: MFCD22199306
CAS Number: 1374659-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR