2-(Difluoromethyl)-3,5-difluoropyridine ≥97%
Supplier: Matrix Scientific
071596-1G
102520-404EA
590.77
USD
102520-404
2-(Difluoromethyl)-3,5-difluoropyridine ≥97%
2-(Difluoromethyl)-3,5-difluoropyridine
Hazard: Irritant
Formula:
C₆H₃F₄N |
MDL Number:
MFCD22199304 CAS Number: 1374659-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...