4-Bromo-3-(difluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

071586-1G
102520-384EA 1223.08 USD
102520-384
4-Bromo-3-(difluoromethyl)pyridine ≥95%
4-Bromo-3-(difluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄BrF₂N
MDL Number: MFCD18257231
CAS Number: 1256821-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR