3-Chloro-5-(difluoromethyl)pyridine ≥95%
Supplier: Matrix Scientific
071582-1G
102520-376EA
895.38
USD
102520-376
3-Chloro-5-(difluoromethyl)pyridine ≥95%
3-Chloro-5-(difluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₄ClF₂N |
MDL Number:
MFCD22199293 CAS Number: 1335055-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...