3,5-Dichloro-4-(difluoromethyl)pyridine ≥95%
Supplier: Matrix Scientific
071581-1G
102520-374EA
412.31
USD
102520-374
3,5-Dichloro-4-(difluoromethyl)pyridine ≥95%
3,5-Dichloro-4-(difluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₃Cl₂F₂N |
MDL Number:
MFCD22124922 CAS Number: 1374659-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...