3-Chloro-4-(difluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

071578-1G
102520-368EA 244.62 USD
102520-368
3-Chloro-4-(difluoromethyl)pyridine ≥97%
3-Chloro-4-(difluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄ClF₂N
MDL Number: MFCD22124921
CAS Number: 1374659-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR