1-Chloro-2,3-bis(trifluoromethyl)benzene ≥95%
Supplier: Matrix Scientific
071566-1G
102520-348EA
396.92
USD
102520-348
1-Chloro-2,3-bis(trifluoromethyl)benzene ≥95%
1-Chloro-2,3-bis(trifluoromethyl)benzene
Hazard: Irritant
Formula:
C₈H₃ClF₆ |
MDL Number:
MFCD16140188 CAS Number: 1221272-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...