1,3-Dichloro-2,4-bis(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

071565-1G
102520-346EA 412.31 USD
102520-346
1,3-Dichloro-2,4-bis(trifluoromethyl)benzene ≥95%
1,3-Dichloro-2,4-bis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₂Cl₂F₆
MDL Number: MFCD16140187
CAS Number: 1221272-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR