1,2,4-Tris(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

071562-1G
102520-338EA 380 USD
102520-338
1,2,4-Tris(trifluoromethyl)benzene ≥95%
1,2,4-Tris(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₃F₉
MDL Number: MFCD16140151
CAS Number: 393-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR