3,5-Dichloro-4-(trifluoromethyl)pyridine ≥95%
Supplier: Matrix Scientific
071557-1G
102520-330EA
381.54
USD
102520-330
3,5-Dichloro-4-(trifluoromethyl)pyridine ≥95%
3,5-Dichloro-4-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₂Cl₂F₃N |
MDL Number:
MFCD16140184 CAS Number: 1221272-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...