1-Methyl-4-(trifluoromethyl)naphthalene ≥95%

Supplier: Matrix Scientific

071552-1G
102520-322EA 684.62 USD
102520-322
1-Methyl-4-(trifluoromethyl)naphthalene ≥95%
1-Methyl-4-(trifluoromethyl)naphthalene

Hazard: Irritant

Formula: C₁₂H₉F₃
MDL Number: MFCD16140174
CAS Number: 52331-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR