2-(Difluoromethyl)naphthalene ≥97%

Supplier: Matrix Scientific

071543-1G
102520-304EA 218.46 USD
102520-304
2-(Difluoromethyl)naphthalene ≥97%
2-(Difluoromethyl)naphthalene

Hazard: Irritant

Formula: C₁₁H₈F₂
MDL Number: MFCD12546940
CAS Number: 140135-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR