1,4-Bis(trifluoromethyl)naphthalene ≥97%

Supplier: Matrix Scientific

071541-1G
102520-300EA 190.77 USD
102520-300
1,4-Bis(trifluoromethyl)naphthalene ≥97%
1,4-Bis(trifluoromethyl)naphthalene

Hazard: Irritant

Formula: C₁₂H₆F₆
MDL Number: MFCD12546938
CAS Number: 52331-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR