3-(Difluoromethyl)quinoline ≥97%

Supplier: Matrix Scientific

071540-1G
102520-298EA 961.54 USD
102520-298
3-(Difluoromethyl)quinoline ≥97%
3-(Difluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₇F₂N
MDL Number: MFCD11226559
CAS Number: 1186195-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR