3-(Difluorophenylmethyl)pyridine ≥95%

Supplier: Matrix Scientific

071539-1G
102520-296EA 283.08 USD
102520-296
3-(Difluorophenylmethyl)pyridine ≥95%
3-(Difluorophenylmethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₉F₂N
MDL Number: MFCD14525566
CAS Number: 1204296-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR