2-(Difluorophenylmethyl)pyridine ≥95%

Supplier: Matrix Scientific

071530-1G
102520-278EA 207.69 USD
102520-278
2-(Difluorophenylmethyl)pyridine ≥95%
2-(Difluorophenylmethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₉F₂N
MDL Number: MFCD14525507
CAS Number: 1204295-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR