1-(1,1-Difluoroethyl)naphthalene ≥95%

Supplier: Matrix Scientific

071526-1G
102520-270EA 235.38 USD
102520-270
1-(1,1-Difluoroethyl)naphthalene ≥95%
1-(1,1-Difluoroethyl)naphthalene

Hazard: Irritant

Formula: C₁₂H₁₀F₂
MDL Number: MFCD14525565
CAS Number: 1204295-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR