1-Bromo-4-(1,1-difluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

071522-1G
102520-264EA 292.31 USD
102520-264
1-Bromo-4-(1,1-difluoropropyl)benzene ≥95%
1-Bromo-4-(1,1-difluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₉BrF₂
MDL Number: MFCD16140148
CAS Number: 162783-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR