1-Bromo-3-(1,1-difluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

071516-1G
102520-252EA 612.31 USD
102520-252
1-Bromo-3-(1,1-difluoropropyl)benzene ≥95%
1-Bromo-3-(1,1-difluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₉BrF₂
MDL Number: MFCD16140163
CAS Number: 1204295-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR