1-(Trifluoromethyl)isoquinoline ≥95%

Supplier: Matrix Scientific

071509-1G
102520-240EA 278.46 USD
102520-240
1-(Trifluoromethyl)isoquinoline ≥95%
1-(Trifluoromethyl)isoquinoline

Hazard: Irritant

Formula: C₁₀H₆F₃N
MDL Number: MFCD11007730
CAS Number: 26053-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR