2-Phenoxybenzotrifluoride ≥95%

Supplier: Matrix Scientific

071501-1G
102520-228EA 267.69 USD
102520-228
2-Phenoxybenzotrifluoride ≥95%
2-Phenoxybenzotrifluoride

Hazard: Irritant

Formula: C₁₃H₉F₃O
MDL Number: MFCD14525537
CAS Number: 106877-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR