1-Methyl-4-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

071500-1G
102520-226EA 229.23 USD
102520-226
1-Methyl-4-(2,2,2-trifluoroethyl)benzene ≥95%
1-Methyl-4-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₉H₉F₃
Melting Pt: 44…46 °C
MDL Number: MFCD14525512
CAS Number: 50562-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR