5-(Trifluoromethyl)-1,2,3,4-tetrahydronaphthalene ≥95%

Supplier: Matrix Scientific

071489-1G
102520-200EA 300 USD
102520-200
5-(Trifluoromethyl)-1,2,3,4-tetrahydronaphthalene ≥95%
5-(Trifluoromethyl)-1,2,3,4-tetrahydronaphthalene

Hazard: Irritant

Formula: C₁₁H₁₁F₃
MDL Number: MFCD14525535
CAS Number: 1204295-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR