2,2,4,4-Tetrafluoropentane ≥97%

Supplier: Matrix Scientific

071470-1G
102520-160EA 126.15 USD
102520-160
2,2,4,4-Tetrafluoropentane ≥97%
2,2,4,4-Tetrafluoropentane

Hazard: Irritant

Formula: C₅H₈F₄
MDL Number: MFCD12546927
CAS Number: 86154-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR