1-(1,1-Difluoropropyl)-4-fluorobenzene ≥95%

Supplier: Matrix Scientific

071453-1G
102520-120EA 267.69 USD
102520-120
1-(1,1-Difluoropropyl)-4-fluorobenzene ≥95%
1-(1,1-Difluoropropyl)-4-fluorobenzene

Hazard: Irritant

Formula: C₉H₉F₃
Boiling Pt: 169 °C
Density: 1.126
Flash Pt: 48 °C
MDL Number: MFCD14525551
CAS Number: 1204295-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR