8-(Benzyloxy)-3-chloroimidazo[1,2-a]pyridine ≥97%
Supplier: Matrix Scientific
071420-500MG
071420-1G
102520-002EA
376.92
USD
102520-002
102520-000
8-(Benzyloxy)-3-chloroimidazo[1,2-a]pyridine ≥97%
8-(Benzyloxy)-3-chloroimidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₁₄H₁₁ClN₂O Melting Pt: 115…117 °C |
MDL Number:
MFCD22126119 CAS Number: 1373350-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...