4-{1H-Imidazo[4,5-b]pyridin-1-yl}piperidine hydrochloride ≥97%
Supplier: Matrix Scientific
071351-500MG
102519-850EA
995.38
USD
102519-850
4-{1H-Imidazo[4,5-b]pyridin-1-yl}piperidine hydrochloride ≥97%
4-{1H-Imidazo[4,5-b]pyridin-1-yl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₄ Melting Pt: 320…324 °C |
MDL Number:
MFCD22126107 CAS Number: 1370587-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...