tert-Butyl (5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-4-yl)carbamate

Supplier: Matrix Scientific

071056-250MG
102519-400EA 709.23 USD
102519-400
tert-Butyl (5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-4-yl)carbamate
tert-Butyl (5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-4-yl)carbamate

Hazard: Irritant

Formula: C₁₃H₁₄F₃N₃O₂
MW: 301.27 g/mol
MDL Number: MFCD20487140
CAS Number: 1346447-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR