5-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile

Supplier: Matrix Scientific

071045-250MG
102519-378EA 720 USD
102519-378
5-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile
5-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile

Hazard: Irritant

Formula: C₈H₃FIN₃
MDL Number: MFCD20487094
CAS Number: 1346446-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR