Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-6-carboxylate

Supplier: Matrix Scientific

071035-250MG
102519-358EA 860 USD
102519-358
Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-6-carboxylate
Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-6-carboxylate

Hazard: Irritant

Formula: C₉H₇ClN₂O₂
MDL Number: MFCD20487060
CAS Number: 1305324-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR