5-Chloro-1H-pyrrolo[2,3-b]pyridin-6-ol

Supplier: Matrix Scientific

071034-250MG
102519-356EA 860 USD
102519-356
5-Chloro-1H-pyrrolo[2,3-b]pyridin-6-ol
5-Chloro-1H-pyrrolo[2,3-b]pyridin-6-ol

Hazard: Irritant

Formula: C₇H₅ClN₂O
MDL Number: MFCD12962803
CAS Number: 1190321-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR