4,5-Dichloro-6-iodo-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

071031-250MG
102519-350EA 860 USD
102519-350
4,5-Dichloro-6-iodo-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
4,5-Dichloro-6-iodo-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₆H₂₃Cl₂IN₂Si
MDL Number: MFCD20487048
CAS Number: 1346447-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR