6-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[3,2-b]pyridine

Supplier: Matrix Scientific

071021-250MG
102519-330EA 675.38 USD
102519-330
6-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[3,2-b]pyridine
6-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₃H₉BrN₂O₂S
MDL Number: MFCD20487024
CAS Number: 1305325-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR