2,3,6-Trimethoxyisonicotinaldehyde

Supplier: Matrix Scientific

071006-250MG
102519-300EA 638.46 USD
102519-300
2,3,6-Trimethoxyisonicotinaldehyde
2,3,6-Trimethoxyisonicotinaldehyde

Hazard: Irritant

Formula: C₉H₁₁NO₄
MDL Number: MFCD21609479
CAS Number: 1364917-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR