4-Iodo-2,3,6-trimethoxypyridine

Supplier: Matrix Scientific

071003-500MG
102519-294EA 913.85 USD
102519-294
4-Iodo-2,3,6-trimethoxypyridine
4-Iodo-2,3,6-trimethoxypyridine

Hazard: Irritant

Formula: C₈H₁₀INO₃
MDL Number: MFCD21609476
CAS Number: 1364917-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR