2-Bromo-5-iodo-3-methoxy-1-methylpyridin-4(1H)-one

Supplier: Matrix Scientific

070981-250MG
102519-250EA 860 USD
102519-250
2-Bromo-5-iodo-3-methoxy-1-methylpyridin-4(1H)-one
2-Bromo-5-iodo-3-methoxy-1-methylpyridin-4(1H)-one

Hazard: Irritant

Formula: C₇H₇BrINO₂
MDL Number: MFCD20486995
CAS Number: 1346446-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR