4-Bromo-2-(tert-butyl)-6-iodo-7-(trimethylsilyl)oxazolo[4,5-c]pyridine

Supplier: Matrix Scientific

070935-250MG
102519-158EA 860 USD
102519-158
4-Bromo-2-(tert-butyl)-6-iodo-7-(trimethylsilyl)oxazolo[4,5-c]pyridine
4-Bromo-2-(tert-butyl)-6-iodo-7-(trimethylsilyl)oxazolo[4,5-c]pyridine

Hazard: Irritant

Formula: C₁₃H₁₈BrIN₂OSi
MDL Number: MFCD20487058
CAS Number: 1305324-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR