4-Bromo-2-(tert-butyl)oxazolo[4,5-c]pyridin-7-ol

Supplier: Matrix Scientific

070931-250MG
102519-150EA 860 USD
102519-150
4-Bromo-2-(tert-butyl)oxazolo[4,5-c]pyridin-7-ol
4-Bromo-2-(tert-butyl)oxazolo[4,5-c]pyridin-7-ol

Hazard: Irritant

Formula: C₁₀H₁₁BrN₂O₂
MDL Number: MFCD20487053
CAS Number: 1305324-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR