4-Bromo-2-(tert-butyl)-7-chloro-6-iodooxazolo[4,5-c]pyridine

Supplier: Matrix Scientific

070928-250MG
102519-144EA 860 USD
102519-144
4-Bromo-2-(tert-butyl)-7-chloro-6-iodooxazolo[4,5-c]pyridine
4-Bromo-2-(tert-butyl)-7-chloro-6-iodooxazolo[4,5-c]pyridine

Hazard: Irritant

Formula: C₁₀H₉BrClIN₂O
MDL Number: MFCD20487007
CAS Number: 1305324-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR