4-Bromo-2-(tert-butyl)-6-iodooxazolo[4,5-c]pyridine

Supplier: Matrix Scientific

070925-250MG
102519-138EA 860 USD
102519-138
4-Bromo-2-(tert-butyl)-6-iodooxazolo[4,5-c]pyridine
4-Bromo-2-(tert-butyl)-6-iodooxazolo[4,5-c]pyridine

Hazard: Irritant

Formula: C₁₀H₁₀BrIN₂O
MDL Number: MFCD20487037
CAS Number: 1305325-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR