tert-Butyl-5-(3-hydroxyprop-1-yn-1-yl)-1H-pyrazolo[3,4-b]pyridine-1-carboxylate

Supplier: Matrix Scientific

070912-250MG
102519-112EA 860 USD
102519-112
tert-Butyl-5-(3-hydroxyprop-1-yn-1-yl)-1H-pyrazolo[3,4-b]pyridine-1-carboxylate
tert-Butyl-5-(3-hydroxyprop-1-yn-1-yl)-1H-pyrazolo[3,4-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₅N₃O₃
MW: 273.29 g/mol
MDL Number: MFCD20487057
CAS Number: 1305325-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR