tert-Butyl-5-((trimethylsilyl)ethynyl)-1H-pyrazolo[3,4-b]pyridine-1-carboxylate
Supplier: Matrix Scientific
070911-250MG
102519-110EA
860
USD
102519-110
tert-Butyl-5-((trimethylsilyl)ethynyl)-1H-pyrazolo[3,4-b]pyridine-1-carboxylate
tert-Butyl-5-((trimethylsilyl)ethynyl)-1H-pyrazolo[3,4-b]pyridine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁N₃O₂Si MW: 315.45 g/mol |
MDL Number:
MFCD20487056 CAS Number: 1305325-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...