5-Iodo-1H-pyrazolo[3,4-b]pyridine

Supplier: Matrix Scientific

070907-250MG
102519-102EA 709.23 USD
102519-102
5-Iodo-1H-pyrazolo[3,4-b]pyridine
5-Iodo-1H-pyrazolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₆H₄IN₃
MDL Number: MFCD20487027
CAS Number: 1305324-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR