7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-6-carbaldehyde
Supplier: Matrix Scientific
070903-250MG
102519-094EA
860
USD
102519-094
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-6-carbaldehyde
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-6-carbaldehyde
Hazard: Irritant
Formula:
C₈H₆ClNO₃ MW: 199.59 g/mol |
MDL Number:
MFCD20487101 CAS Number: 1346447-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...