tert-Butyl (7-chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)carbamate

Supplier: Matrix Scientific

070902-250MG
102519-092EA 860 USD
102519-092
tert-Butyl (7-chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)carbamate
tert-Butyl (7-chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)carbamate

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₄
MW: 286.72 g/mol
MDL Number: MFCD20487100
CAS Number: 1346447-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR