tert-Butyl ((7-chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)methyl)carbamate
Supplier: Matrix Scientific
070901-250MG
102519-090EA
612.31
USD
102519-090
tert-Butyl ((7-chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)methyl)carbamate
tert-Butyl ((7-chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)methyl)carbamate
Hazard: Irritant
Formula:
C₁₃H₁₇ClN₂O₄ MW: 300.74 g/mol |
MDL Number:
MFCD20487096 CAS Number: 1346447-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...